Chemical resistant gloves, AlphaTec® 87-950 / 87-955
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
87-955/10.5-11 87-955/7.5-8
112-0309EA 0 DKK
112-0309 112-0311
Chemical resistant gloves, AlphaTec® 87-950 / 87-955
Handsker
Disse bomuldforede og klorerede handsker med en høj svedabsorbans, er komfortable at have på og er påtrykt et rudemønster, som giver et bedre greb.
  • Excellent resistance to mechanical hazards and increased chemical protection
  • Extremely high resistance to many ketones, salts, detergents, alcohols, alkalis and fats
  • Chlorinated: For better grip and increased chemical resistance
Suitable for heavy duty handling, chemical and petrol processing and maintenance activities.
Certificeringer: EN ISO 374-1 Type A (AKLOPS), EN ISO 374-5 VIRUS, EN 388:2016 (X121X); Cat. III