Dibutyltinoxid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide

OR921569-5G OR921569-100G
APOSOR921569-5GEA 164 DKK
APOSOR921569-5G APOSOR921569-100G
Dibutyltinoxid 95%
Dibutyltinoxid
Formel: C₈H₁₈OSn
Smeltepunkt: >300 °C
Massefylde: 1,580
MDL Number: MFCD00001992
CAS-nummer: 818-08-6
UN nummer: 3146
ADR: 6.1,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR