Live or Dead™ Cell Viability Assay Kits *Green/Red Dual Fluorescence*
Leverandør: AAT BIOQUEST
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22760 22789
AATB22760EA 9260 DKK
AATB22760 AATB22789
Live or Dead™ Cell Viability Assay Kits *Green/Red Dual Fluorescence*
Assays Cellulær Assays
This Live or Dead™cell viability uses two fluorogenic indicators: calcein AM for viable cells and a cell-impermeable DNA-binding dye for the cells with compromised membranes.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
  • Results in under 2 hours
Advarsel: For research use only.