Genomic DNA ScreenTape Analysis (ScreenTape and Reagents)
Leverandør: AGILENT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5067-5365 5067-5366
732-3995EA 2210 DKK
732-3995 732-3996
Genomic DNA ScreenTape Analysis (ScreenTape and Reagents)
Elektroforesesystemer Kapillær Elektroforesesystemer
Integrity assessment of genomic DNA samples up to greater than 60000 bp.
  • DNA Integrity Number (DIN) is used for accurate and objective assessments of genomic DNA degradation
  • Results may be obtained in less than 2 minutes per sample
  • Input sample QC allows saving of sequencing and sample preparation in Next Generation Sequencing workflows
  • Ready-to-use ScreenTape devices and automated data analysis provide an easy-to-use system with excellent reproducibility
  • Partially used ScreenTape devices can be used for completion at a later point in time
  • No cross-contamination or sample carryover is caused due to the use of single lanes and individual loading tips
Genomic DNA ScreenTape analysis provides fast, automated and reliable electrophoretic separation of input genomic DNA samples. The gDNA ScreenTape assay can separate DNA samples from 200 to more than 60000 base pairs in less than 2 minutes/sample. Samples can be analysed from multiple sources including FFPE.
Objective gDNA integrity assessment is delivered with a numeric measurement, the DNA Integrity Number (DIN).
The assay is compatible with all Agilent TapeStation instruments.
Note: Some products within this assay contain SYBR™ Green I, which is licensed from Life Technologies Corporation for use in research and development only. SYBR™ is a registered trademark of Life Technologies Corporation.
Advarsel: For research use only. Not for use in diagnostic procedures.