1-[2-Nitro-4-(trifluoromethyl)phenyl]-3-methylpiperazine hydrochloride
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
3-Methyl-1-[2-nitro-4-(trifluoromethyl)phenyl]piperazine hydrochloride
PC7456-1G
PC7456-5G
APOSPC7456-1GEA
830
DKK
APOSPC7456-1G
APOSPC7456-5G
1-[2-Nitro-4-(trifluoromethyl)phenyl]-3-methylpiperazine hydrochloride
1-[2-Nitro-4-(trifluoromethyl)phenyl]-3-methylpiperazine hydrochloride
Formel:
C₁₂H₁₅ClF₃N₃O₂ Smeltepunkt: 159…161 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01446115 CAS-nummer: 299426-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...