Balsalazide disodium salt dihydrate

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-[4-(2-Carboxyethylcarbamoyl)phenylazo]salicylic Acid Disodium Salt, Sodium (E)-5-((4-((2-carboxylatoethyl)carbamoyl)phenyl)diazenyl)-2-hydroxybenzoate dihydrate

465090010 465090050
ACRO465090010EA 400 DKK
ACRO465090010 ACRO465090050
Balsalazide disodium salt dihydrate
Balsalazide disodium salt dihydrate

  • Bioactive

Formel: C₁₇H₁₇N₃Na₂O₈
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01631117
CAS-nummer: 150399-21-6

Bestil nu

Specifikationer Testresultater

Appearance Yellow to yellow-red Crystalline powder
Infrared spectrum Conforms
Water 7 to 10 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR