Thiobencarb (S-4-Chlorbenzyl diethylthiocarbamat) ≥97.0% (by GC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Thiobencarb, S-(4-Chlorobenzyl) diethylcarbamothioate, Benthiocarb, Diethylthiocarbamic acid S-(4-Chlorobenzyl) ester

T3881-1G T3881-5G
TCIAT3881-1GEA 698 DKK
TCIAT3881-1G TCIAT3881-5G
Thiobencarb (S-4-Chlorbenzyl diethylthiocarbamat) ≥97.0% (by GC, total nitrogen)
Thiobencarb (S-4-Chlorbenzyl diethylthiocarbamat)
Formel: C₁₂H₁₆ClNOS
Kogepunkt: 131°C
Smeltepunkt: 3°C
MDL Number: MFCD00055475
CAS-nummer: 28249-77-6
EINECS: 248-924-5
UN nummer: 3082
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 97.0 %
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR