3-(2-Methoxyphenyl)propionsyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-(2-Methoxyphenyl)propanoic acid
M2673-5G
M2673-25G
TCIAM2673-5GEA
411
DKK
TCIAM2673-5G
TCIAM2673-25G
3-(2-Methoxyphenyl)propionsyre ≥98.0% (by GC, titration analysis)
3-(2-Methoxyphenyl)propionsyre
Formel:
C₁₀H₁₂O₃ |
MDL Number:
MFCD00002772 CAS-nummer: 6342-77-4 EINECS: 228-738-0 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 86.0 to 90.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...