3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3,4-Difluoro-4'-(trans-4-ethylcyclohexyl)biphenyl

D4859-5G D4859-25G
TCIAD4859-5GEA 485 DKK
TCIAD4859-5G TCIAD4859-25G
3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl ≥98.0% (by GC)
3,4-Difluoro-4'-(4-ethylcyclohexyl)biphenyl
Formel: C₂₀H₂₂F₂
MDL Number: MFCD12828121
CAS-nummer: 134412-18-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR