4,4'-Dihydroxyazobenzene ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4,4-Dihydroxyazobenzene, 4,4'-Azodiphenol
D3431-1G
D3431-5G
TCIAD3431-1GEA
772
DKK
TCIAD3431-1G
TCIAD3431-5G
4,4'-Dihydroxyazobenzene ≥98.0% (by HPLC, titration analysis)
4,4'-Dihydroxyazobenzene
Formel:
C₁₂H₁₀N₂O₂ |
MDL Number:
MFCD00045778 CAS-nummer: 2050-16-0 |
Specifikationer Testresultater
Appearance | Amber to Brown to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...