Carvedilol ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol

C2260-25G C2260-5G
TCIAC2260-25GEA 2570 DKK
TCIAC2260-25G TCIAC2260-5G
Carvedilol ≥98.0% (by HPLC, titration analysis)
Carvedilol
Formel: C₂₄H₂₆N₂O₄
MDL Number: MFCD00864692
CAS-nummer: 72956-09-3
UN nummer: 3077
ADR: 9,III
Merck Index: 14,01873
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89cd7bf623930b02fa6f9e363654898b8797ff30&vid=89cd7bf623930b02fa6f9e363654898b8797ff30&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR