Chloromethylstyrene (m- and p- mixture) ≥88.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Chloromethylstyrene, Vinylbenzyl Chloride (m and p mixture)
Stabilization:
Stabilised with TBC + ONP + o-nitrocresol
C0767-25G
C0767-500G
TCIAC0767-25GEA
263
DKK
TCIAC0767-25G
TCIAC0767-500G
Chloromethylstyrene (m- and p- mixture) ≥88.0% (by GC) stabilised
Chloromethylstyrene (m and p mixture)
Formel:
C₉H₉Cl Flammepunkt: 107°C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00051362 CAS-nummer: 57458-41-0 UN nummer: 3265 ADR: 8,III |
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 88.0 % (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...