Sodium-2-chlorotoluene-4-sulphonate ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Sodium 3-Chloro-4-methylbenzenesulphonate, 2-Chlorotoluene-4-sulphonic acid Sodium Salt

C0298-25G C0298-500G
TCIAC0298-25GEA 255 DKK
TCIAC0298-25G TCIAC0298-500G
Sodium-2-chlorotoluene-4-sulphonate ≥95.0% (by titrimetric analysis)
Sodium-2-chlorotoluene-4-sulphonate
Formel: C₇H₇ClO₃S
MDL Number: MFCD00053619
CAS-nummer: 5138-91-0
EINECS: 225-901-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 95.0 %
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR