Benzylbis(triphenylphosphine)palladium(II) chloride ≥96.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (SP-4-3)-Benzylbis(triphenylphosphine)palladium(II) chloride, trans-Benzyl(chloro)bis(triphenylphosphine)palladium(II), Benzyl(chloro)bis(triphenylphosphine)palladium(II)

B2029-1G B2029-100MG
TCIAB2029-1GEA 1430 DKK
TCIAB2029-1G TCIAB2029-100MG
Benzylbis(triphenylphosphine)palladium(II) chloride ≥96.0% (by titrimetric analysis)
Benzylbis(triphenylphosphine)palladium(II) chloride
Formel: C₄₃H₃₇ClP₂Pd
Opbevaringstemperatur: Fryser
MDL Number: MFCD00010395
CAS-nummer: 22784-59-4

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Chelometric Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR