Heptafluorsmørsyre ∼0.5 mol/L in water for ion pair chromatography, for LC-MS
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid
A5713-10ML
A5713-100ML
TCIAA5713-10MLEA
247
DKK
TCIAA5713-10ML
TCIAA5713-100ML
Heptafluorsmørsyre ∼0.5 mol/L in water for ion pair chromatography, for LC-MS
Heptafluorsmørsyre
Formel:
C₄HF₇O₂ |
MDL Number:
MFCD00004171 CAS-nummer: 375-22-4 EINECS: 206-786-3 UN nummer: 3265 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | Colorless clear liquid |
Concentration (Neutralization titration) | 0.50 to 0.55 mol/L |
Absorbance (254nm) | max. 0.350 |
Absorbance (270nm) | max. 0.100 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...