2-Phenoxyethyl acetate ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Acetic Acid 2-Phenoxyethyl Ester, Ethylene Glycol Monophenyl Ether Acetate

A0935-25G A0935-500G
TCIAA0935-25GEA 181 DKK
TCIAA0935-25G TCIAA0935-500G
2-Phenoxyethyl acetate ≥99.0% (by GC)
2-Phenoxyethyl acetate
Formel: C₁₀H₁₂O₃
Flammepunkt: 141°C
MDL Number: MFCD00059336
CAS-nummer: 6192-44-5

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR