4-Brom-3-Fluoranilin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B22227.09 B22227.88
B22227.09EA 1610 DKK
B22227.09 B22227.88
4-Brom-3-Fluoranilin ≥98%
4-Brom-3-Fluoranilin

Purity: 98% CAS No.: 656-65-5 Molecular Formula: C6H5BrFN Formula Weight: 190.01 Flash Point: 104°(219°F) Melting Point: 65-70° Beilstein Reference: 3236457 UN No.: 2811 MDL No.: MFCD00672933

Formel: C₆H₅BrFN
Kogepunkt: 231 °C (1013 hPa)
Smeltepunkt: 65…70 °C
Flammepunkt: 104 °C (219 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00672933
CAS-nummer: 656-65-5
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR