3-(Dimethylamino)acrolein 90%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-(Dimethylamino)acrylaldehyde, 3-(Dimethylamino)propenal, 3-(Dimethylamino)-2-propenal

433320050 433320250
ACRO433320050EA 508 DKK
ACRO433320050 ACRO433320250
3-(Dimethylamino)acrolein 90%
3-(Dimethylamino)acrolein
Formel: C₅H₉NO
Kogepunkt: 270…273 °C (760 mmHg)
Massefylde: 0,77 g/cm³
Flammepunkt: 113 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00006999
CAS-nummer: 927-63-9
UN nummer: 3267

Bestil nu

Specifikationer Testresultater

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
GC ≥99.0 %
Water ≤0.5 %
UV at 310 nm A: ≤0.01
Relative density (20/20°C) 0.785 to 0.789
Specific gravity (25°C) 0.783 to 0.787
Identification D Passes test
Non-volatile matter ≤20 ppm
Related substances ≤0.3 %
Acidity/alkalinity Passes test
Benzene ≤2 ppm
Peroxides Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR