4-Isocyanatobenzeneboronic acid pinacol ester 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H53003.MD
H53003.03
H53003.MDEA
811
DKK
H53003.MD
H53003.03
4-Isocyanatobenzeneboronic acid pinacol ester 95%
4-Isocyanatobenzeneboronic acid pinacol ester
Purity: 95%
CAS No.: 380430-64-8
Molecular Formula: C13H16BNO3
Formula Weight: 245.08
Melting Point: 162-168°
MDL No.: MFCD03411942
Formel:
C₁₃H₁₆BNO₃ Smeltepunkt: 162…168 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03411942 CAS-nummer: 380430-64-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...