5-Nitro-2-picoline ≥96.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Methyl-5-nitropyridine

M2034-1G M2034-5G
TCIAM2034-1GEA 460 DKK
TCIAM2034-1G TCIAM2034-5G
5-Nitro-2-picoline ≥96.0% (by GC, titration analysis)
5-Nitro-2-picoline
Formel: C₆H₆N₂O₂
MDL Number: MFCD04114179
CAS-nummer: 21203-68-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa5a552db9af174a7a17bda77807bf289d74b432&vid=aa5a552db9af174a7a17bda77807bf289d74b432&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 107.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR