Isoquinoline-5-sulphonic acid ≥96.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

I0470-25G I0470-250G
TCIAI0470-25GEA 345 DKK
TCIAI0470-25G TCIAI0470-250G
Isoquinoline-5-sulphonic acid ≥96.0% (by HPLC, titration analysis)
Isoquinoline-5-sulphonic acid
Formel: C₉H₇NO₃S
MDL Number: MFCD00134089
CAS-nummer: 27655-40-9
UN nummer: 2585
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45a47bbefee73047f6738bbd9a24c7bc44e8e4ae&vid=45a47bbefee73047f6738bbd9a24c7bc44e8e4ae&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR