4-Nitrobenzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (4-Nitrophenyl)methanamine hydrochloride

N0435-25G N0435-5G
TCIAN0435-25GEA 7320 DKK
TCIAN0435-25G TCIAN0435-5G
4-Nitrobenzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Nitrobenzylamine hydrochloride
Formel: C₇H₉ClN₂O₂
MDL Number: MFCD00012863
CAS-nummer: 18600-42-5
EINECS: 242-441-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7e7207a97d72e1d502a64185321cb105804db8f&vid=a7e7207a97d72e1d502a64185321cb105804db8f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR