Phenyl acrylate ≥98.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Stabilization: stabiliseret med BHT/Ionol

P2268-5G P2268-25G
TCIAP2268-5GEA 370 DKK
TCIAP2268-5G TCIAP2268-25G
Phenyl acrylate ≥98.0% (by GC) stabilised
Phenyl acrylate
Formel: C₉H₈O₂
Kogepunkt: 98°C
MDL Number: MFCD00048145
CAS-nummer: 937-41-7
EINECS: 213-329-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72f6dd42745c6bc96da1304c63bb3a70b1bf0575&vid=72f6dd42745c6bc96da1304c63bb3a70b1bf0575&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR