1,3-Bis(brommethyl)benzen ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,3-Bis(bromomethyl)benzene, m-Xylylene dibromide

D0215-25G D0215-5G
TCIAD0215-25GEA 1510 DKK
TCIAD0215-25G TCIAD0215-5G
1,3-Bis(brommethyl)benzen ≥97.0% (by GC)
1,3-Bis(brommethyl)benzen
Formel: C₈H₈Br₂
Kogepunkt: 140°C
Smeltepunkt: 76…78 °C
Massefylde: 1,959 g/cm³ (0 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000178
CAS-nummer: 626-15-3
EINECS: 210-931-6
UN nummer: 2928
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61f8a37347880283461dabc963c34f8d7997d371&vid=543d7d82031b49b11e993db0abeaf94a18bcbce2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 75.0 to 78.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR