m-Cresol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: m-Methylphenol

C0401-25G C0401-500G
TCIAC0401-25GEA 132 DKK
TCIAC0401-25G TCIAC0401-500G
m-Cresol ≥98.0% (by GC)
m-Cresol
Formel: CH₃C₆H₄OH
Kogepunkt: 202°C
Massefylde: 1,0336 g/cm³ (20 °C)
Flammepunkt: 86°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002302
CAS-nummer: 108-39-4
EINECS: 203-577-9
UN nummer: 2076
ADR: 6.1,II
Merck Index: 14,02579
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c5b5933636a8d0c884930b6ecd5361c550d240c&vid=8c5b5933636a8d0c884930b6ecd5361c550d240c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR