Methylthiomethyl acetate ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Acetic Acid Methylthiomethyl Ester

A1368-25G A1368-5G
TCIAA1368-25GEA 698 DKK
TCIAA1368-25G TCIAA1368-5G
Methylthiomethyl acetate ≥95.0% (by GC)
Methylthiomethyl acetate
Formel: C₄H₈O₂S
Kogepunkt: 62°C
Flammepunkt: 44°C
MDL Number: MFCD00075528
CAS-nummer: 16437-69-7
UN nummer: 3272
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR