Aluminium tri-tert-butanolat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Aluminium tert-butoxide, Aluminumtert-butoxide

A0804-5G
TCIAA0804-5GEA 329 DKK
TCIAA0804-5G
Aluminium tri-tert-butanolat ≥98.0% (by titrimetric analysis)
Aluminium tri-tert-butanolat
Formel: C₁₂H₂₇AlO₃
MDL Number: MFCD00008801
CAS-nummer: 556-91-2
EINECS: 209-146-1
UN nummer: 2925
ADR: 4.1,III
Merck Index: 14,00333

Bestil nu

Specifikationer Testresultater

Appearance White to Gray powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR