Detergents for Glassware Washers
Leverandør: SMEG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DETERLQC2 ACIDGLP2 ACIDGLC2 DETERLQSP2 DETERLIQUIDI2
SMEGDETERLQC2EA 873 DKK
SMEGDETERLQC2 SMEGACIDGLP2 SMEGACIDGLC2 SMEGDETERLQSP2 SMEGDETERLIQUIDI2
Detergents for Glassware Washers
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Detergents designed to optimise cleaning efficiency and the thermal disinfection phase for laboratory glassware cleaning.
  • For thorough instrument cleaning
  • Alkaline detergents for washing and neutralisers for the neutralisation stage
DETERGLASS
• Universal alkaline powder
• Detergent for automatic washing of laboratory glassware
• Contains sodium hydroxide, sodium metasilicate pentahydrate, sodium triphosphate pentabasic, triclosene sodium dihydrate
• 10 kg pack
DETERLIQUID C2
• Strong universal liquid alkaline detergent phosphate-free for washing steelMore Product Information