Tri(2-furyl)phosphine ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tri(furan-2-yl)phosphine, Tri-2-furylphosphine

L13329.03 L13329.06 L13329.MD
L13329.03EA 982 DKK
L13329.03 L13329.06 L13329.MD
Tri(2-furyl)phosphine ≥97%
Tri(2-furyl)phosphine

Purity: 97% CAS No.: 5518-52-5 Molecular Formula: C12H9O3P Formula Weight: 232.18 Melting Point: 59-64° Boiling Point: 136°/4mm MDL No.: MFCD00151857

Formel: C₁₂H₉O₃P
Kogepunkt: 136 °C (4 mmHg)
Smeltepunkt: 59…64 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00151857
CAS-nummer: 5518-52-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR