3,5-Bis(trifluoromethyl)phenyl Isocyanate 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1-Isocyanato-3,5-bis(trifluoromethyl)benzene, 3,5-Bis(trifluoromethyl)phenylisocyanate, Isocyanic Acid 3,5-Bis(trifluoromethyl)phenyl Ester

L09582.03 L09582.06
L09582.03EA 459 DKK
L09582.03 L09582.06
3,5-Bis(trifluoromethyl)phenyl Isocyanate 98%
3,5-Bis(trifluoromethyl)phenyl Isocyanate

Purity: 98% CAS No.: 16588-74-2 Molecular Formula: C9H3F6NO Formula Weight: 255.12 Storage Temperature: 2°C to 8°C Flash Point: 67°(152°F) Refractive Index: 1.4300 Odour: Pungent Boiling Point: 62°/2mm Beilstein Reference: 542272 UN No.: UN2206 MDL No.: MFCD00013559

Formel: C₉H₃F₆NO
Kogepunkt: 62 °C (2 mmHg)
Massefylde: 1,476
Flammepunkt: 67 °C (152 °F)
MDL Number: MFCD00013559
CAS-nummer: 16588-74-2
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR