4-Nitro-o-phenylenediamin 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Nitro-1,2-phenylenediamine, 4-Nitrobenzene-1,2-diamine

L04693.22 L04693.36
L04693.22EA 913 DKK
L04693.22 L04693.36
4-Nitro-o-phenylenediamin 97%
4-Nitro-o-phenylenediamin

Purity: 97% CAS No.: 99-56-9 Molecular Formula: C6H7N3O2 Formula Weight: 153.14 Melting Point: 199-203° Beilstein Reference: 608106 Merck Reference: 14,6623 MDL No.: MFCD00007724

Formel: C₆H₇N₃O₂
Smeltepunkt: 199…203 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007724
CAS-nummer: 99-56-9
EINECS: 202-766-3
Merck Index: 14,6623

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR