Anti-DAPK1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverandør: Bioss
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BS-1687R-A488
BOSSBS-1687R-A488EA 6730 DKK
BOSSBS-1687R-A488
Anti-DAPK1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Calcium/calmodulin-dependent serine/threonine kinase involved in multiple cellular signaling pathways that trigger cell survival, apoptosis, and autophagy. Regulates both type I apoptotic and type II autophagic cell deaths signal, depending on the cellular setting. The former is caspase-dependent, while the latter is caspase-independent and is characterized by the accumulation of autophagic vesicles. Phosphorylates PIN1 resulting in inhibition of its catalytic activity, nuclear localization, and cellular function. Phosphorylates TPM1, enhancing stress fiber formation in endothelial cells.More Product Information