Tetrabutylammoniumhexafluorphosphat 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate

PC6634-25G PC6634-100G PC6634-500G
APOSPC6634-25GEA 240 DKK
APOSPC6634-25G APOSPC6634-100G APOSPC6634-500G
Tetrabutylammoniumhexafluorphosphat 98%
Tetrabutylammoniumhexafluorphosphat
Formel: C₁₆H₃₆F₆NP
Smeltepunkt: 244…248 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011748
CAS-nummer: 3109-63-5
EINECS: 221-472-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR