Methylindol-1-carboxylat 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 1H-indole-1-carboxylate

OR933004-1G OR933004-5G
APOSOR933004-1GEA 1430 DKK
APOSOR933004-1G APOSOR933004-5G
Methylindol-1-carboxylat 95%
Methylindol-1-carboxylat
Formel: C₁₀H₉NO₂
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 39203-20-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR