Diethoxy(dimethyl)silan

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dimethyldiethoxysilane

OR61048-100G OR61048-2.5KG OR61048-500G
APOSOR61048-100GEA 164 DKK
APOSOR61048-100G APOSOR61048-2.5KG APOSOR61048-500G
Diethoxy(dimethyl)silan
Diethoxy(dimethyl)silan
Formel: C₆H₁₆O₂Si
Kogepunkt: 111…113 °C
Smeltepunkt: –70 °C
Massefylde: 0,865 g/cm³ (25 °C)
Flammepunkt: 11 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009068
CAS-nummer: 78-62-6
EINECS: 201-127-6
UN nummer: 2380
ADR: 3,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR