3-Morpholineethanol hydrochloride 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR452075-100MG OR452075-250MG OR452075-500MG
APOSOR452075-100MGEA 2460 DKK
APOSOR452075-100MG APOSOR452075-250MG APOSOR452075-500MG
3-Morpholineethanol hydrochloride 97%
3-Morpholineethanol hydrochloride
Formel: C₆H₁₃NO₂·ClH
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22199183
CAS-nummer: 399580-64-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR