Methyl-4-azaindole-2-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR41118-1G OR41118-250MG
APOSOR41118-1GEA 3550 DKK
APOSOR41118-1G APOSOR41118-250MG
Methyl-4-azaindole-2-carboxylate
Methyl-4-azaindole-2-carboxylate
Formel: C₉H₈N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09743416
CAS-nummer: 394223-19-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR