tert-Butyl-3-hydroxycyclobutylcarbamate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: tert-butyl N-(3-hydroxycyclobutyl)carbamate, tert-Butyl (3-hydroxycyclobutyl)carbamate

OR301347-1G OR301347-250MG
APOSOR301347-1GEA 218 DKK
APOSOR301347-1G APOSOR301347-250MG
tert-Butyl-3-hydroxycyclobutylcarbamate
tert-Butyl-3-hydroxycyclobutylcarbamate
Formel: C₉H₁₇NO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11501030
CAS-nummer: 154748-63-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR