Jern (III) acetylacetonat

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

OR15745-100G OR15745-500G
APOSOR15745-100GEA 491 DKK
APOSOR15745-100G APOSOR15745-500G
Jern (III) acetylacetonat
Jern (III) acetylacetonat
Formel: C₁₅H₂₁FeO₆
Smeltepunkt: 180…182 °C
Massefylde: 5,24 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000020
CAS-nummer: 14024-18-1
EINECS: 237-853-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR