Ethyl-5-hydroxy-2-methyl-1H-indole-3-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Hydroxy-2-methylindole-3-carboxylic acid ethyl ester, Ethyl 5-hydroxy-2-methyl-1H-indole-3-carboxylate

BIB6068-25G
APOSBIB6068-25GEA 5100 DKK
APOSBIB6068-25G
Ethyl-5-hydroxy-2-methyl-1H-indole-3-carboxylate
Ethyl-5-hydroxy-2-methyl-1H-indole-3-carboxylate
Formel: C₁₂H₁₃NO₃
Smeltepunkt: 205…208 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00044575
CAS-nummer: 7598-91-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR