2-Phenyl-5-benzimidazolesulphonic acid 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ensulizole, Phenylbenzimidazole sulphonic acid, 1H-Benzimidazole-5-sulphonic acid

259700250 259700050 259702500
ACRO259700250EA 368 DKK
ACRO259700250 ACRO259700050 ACRO259702500
2-Phenyl-5-benzimidazolesulphonic acid 98%
2-Phenyl-5-benzimidazolesulphonic acid

CAS No.: 27503-81-7

Formel: C₁₃H₁₀N₂O₃S
Smeltepunkt: >300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00053007
CAS-nummer: 27503-81-7

Bestil nu

Specifikationer Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤7 %
Titration excess NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR