Iminodiacetic acid for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,2'-Azanediyldiacetic acid

M-Clarity™ quality level = MQ200

8.14165.0250
8.14165.0250EA 288 DKK
8.14165.0250
Iminodiacetic acid for synthesis, Sigma-Aldrich®
Iminodiacetic acid
Formel: C₄H₇NO₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004280
CAS-nummer: 142-73-4
EINECS: 205-555-4

Bestil nu

Specifikationer Testresultater

Solubility 42 g/l (20 °C)
Melting Point 247 °C (decomposition)
Molar Mass 133.10 g/mol
pH 2.2 - 2.3 (H2O, 20 °C) (saturated solution)
Hill Formula C4H7NO4
Chemical Formula HN(CH2COOH)2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR