Indium(III)acetylacetonat ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Indium (III) tris(2,4-pentanedionato-O,O'), Indium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')indium (III)

13794.03 13794.06 13794.14
13794.03EA 571 DKK
13794.03 13794.06 13794.14
Indium(III)acetylacetonat ≥98%
Indium(III)acetylacetonat

Purity: 98% CAS No.: 14405-45-9 Molecular Formula: C15H21InO6 Formula Weight: 412.15 Physical Form: Powder Melting Point: 186° dec. MDL No.: MFCD00013494


Used in catalysts

Formel: C₁₅H₂₁InO₆
Smeltepunkt: 186 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013494
CAS-nummer: 14405-45-9
EINECS: 238-378-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR